This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other HSD17B7 SIRNA Assays ›
Gene Symbol:
Hsd17b7
Gene Name:
hydroxysteroid (17-beta) dehydrogenase 7
Gene Aliases:
-
Chromosome Location:
Chr.13: 88288116 - 88308011 on Build Rnor_6.0
UniGene ID:
Rn.7040
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID s131662

Purification
Size
Availability Made To Order
Catalog # 4390771
Price 368.00
Your Price
Online offer: 309.12
309.1
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Hsd17b7
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_017235.3 NP_058931.1 4 463
XM_006250215.2 XP_006250277.1 4 486
XM_006250216.2 XP_006250278.1 4 486
XM_006250217.2 XP_006250279.1 4 486
GenBank mRNA FM073997.1 - 4 456
FN803430.1 - 2 76
FQ113345.1 - 4 463
U44803.1 - 4 408

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

cholesterol biosynthetic process
estrogen biosynthetic process
oxidation-reduction process
3-keto sterol reductase activity
estradiol 17-beta-dehydrogenase activity
prolactin receptor binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0