This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other PADI2 SIRNA Assays ›
Gene Symbol:
Padi2
Gene Name:
peptidyl arginine deiminase, type II
Gene Aliases:
Pdi2
Chromosome Location:
Chr.5: 159428491 - 159471113 on Build Rnor_6.0
UniGene ID:
Rn.98523
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID s131597

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Padi2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_017226.1 NP_058922.1 16 1840
XM_008764215.1 XP_008762437.1 17 1944
GenBank mRNA J05022.1 - 16 1840

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_008812, AK033760, AK158382, AK148156, AK154814, and others.

Gene Ontology Categories:

Function(s) Process(es)

regulation of chromatin disassembly
protein citrullination
substantia nigra development
intracellular estrogen receptor signaling pathway
histone H3-R26 citrullination
chromatin-mediated maintenance of transcription
negative regulation of chemokine-mediated signaling pathway
negative regulation of lymphocyte chemotaxis
protein-arginine deiminase activity
calcium ion binding
estrogen receptor binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0