This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other SLC22A2 SIRNA Assays ›
Gene Symbol:
Slc22a2
Gene Name:
solute carrier family 22 (organic cation transporter), member 2
Gene Aliases:
OCT2, OCT2r, rOCT2
Chromosome Location:
Chr.1: 48318025 - 48360219 on Build Rnor_6.0
UniGene ID:
Rn.10555
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID s131579

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Slc22a2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_031584.2 NP_113772.1 10 1632
GenBank mRNA D83044.1 - 10 1635
X98334.1 - 10 1581
Y13154.1 - 10 1582

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

cation transport
organic cation transport
quaternary ammonium group transport
choline transport
ion transmembrane transport
histamine transport
cation transmembrane transport
steroid binding
ion transmembrane transporter activity
organic cation transmembrane transporter activity
choline transmembrane transporter activity
quaternary ammonium group transmembrane transporter activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0