This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other AES SIRNA Assays ›
Gene Symbol:
Aes
Gene Name:
amino-terminal enhancer of split
Gene Aliases:
Grg, Grg-5
Chromosome Location:
Chr.7: 11012843 - 11018156 on Build Rnor_6.0
UniGene ID:
Rn.11495
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID s131482

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Aes
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_019220.1 NP_062093.1 2, 3 267
GenBank mRNA BC101855.1 - 232 7933368
L14462.1 - 2, 3 267

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

negative regulation of transcription from RNA polymerase II promoter
skeletal system development
transcription, DNA-templated
regulation of transcription from RNA polymerase II promoter
negative regulation of gene expression
Wnt signaling pathway
cellular response to extracellular stimulus
negative regulation of protein binding
regulation of growth
negative regulation of transcription, DNA-templated
negative regulation of response to cytokine stimulus
response to interleukin-1
negative regulation of canonical Wnt signaling pathway
positive regulation of anoikis
transcription corepressor activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0