This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other AVPR1B SIRNA Assays ›
Gene Symbol:
Avpr1b LOC100909648
Gene Name:
arginine vasopressin receptor 1B
vasopressin V1b receptor-like
Gene Aliases:
-
-
Chromosome Location:
Chr.13: 48367307 - 48379134 on Build Rnor_6.0
Chromosome Location:
Chr.13: 48390434 - 48400768 on Build Rnor_6.0
UniGene ID:
Rn.10096
-
Species:
Rat
Species Specific ID (Flybase ID):
-
-
siRNA ID s131470

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Avpr1b
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_017205.3 NP_058901.3 2 1330
XM_008769445.1 XP_008767667.1 1 990
GenBank mRNA D45400.1 - 1 847
U27322.1 - 1 1223
Gene Symbol: LOC100909648
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001289800.1 NP_001276729.1 1 798
XM_008769404.1 XP_008767626.1 1 983

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_011924, AK050523, AK033416.

Gene Ontology Categories:

Function(s) Process(es)

regulation of systemic arterial blood pressure by vasopressin
G-protein coupled receptor signaling pathway
adenylate cyclase-modulating G-protein coupled receptor signaling pathway
positive regulation of cytosolic calcium ion concentration
positive regulation of glutamate secretion
positive regulation of cellular pH reduction
cellular response to hormone stimulus
hyperosmotic salinity response
positive regulation of vasoconstriction
response to peptide
G-protein coupled receptor activity
vasopressin receptor activity
peptide hormone binding
protein homodimerization activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-7db4fb6694-9hb2s:80/100.66.128.142:80.
git-commit: 672a5a8c50d6b616e949be631a941c86748ade1b
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header