This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other MECP2 SIRNA Assays ›
Gene Symbol:
Mecp2
Gene Name:
methyl CpG binding protein 2
Gene Aliases:
-
Chromosome Location:
Chr.X: 156650389 - 156713813 on Build Rnor_6.0
UniGene ID:
Rn.38416
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID s131300

Purification
Size
Availability Made To Order
Catalog # 4390771
Price 368.00
Your Price
Online offer: 309.12
309.1
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Mecp2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_022673.2 NP_073164.2 3 345
XM_006229566.2 XP_006229628.1 3 519
XM_006229567.2 XP_006229629.1 2 629
GenBank mRNA DQ897368.1 - 2 221
DQ897369.1 - 3 345
M94064.1 - 2 187

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

negative regulation of transcription from RNA polymerase II promoter
behavioral fear response
response to hypoxia
startle response
neurological system process involved in regulation of systemic arterial blood pressure
regulation of respiratory gaseous exchange by neurological system process
inositol metabolic process
mitochondrial electron transport, ubiquinol to cytochrome c
chromatin silencing
regulation of gene expression by genetic imprinting
transcription, DNA-templated
regulation of transcription, DNA-templated
glutamine metabolic process
cellular biogenic amine metabolic process
chemical synaptic transmission
synapse assembly
brain development
heart development
respiratory gaseous exchange
learning
memory
long-term memory
protein localization
glucocorticoid metabolic process
positive regulation of cell proliferation
adult locomotory behavior
visual learning
pathogenesis
post-embryonic development
response to ionizing radiation
response to lead ion
regulation of gene expression
response to organic cyclic compound
dendrite development
histone methylation
histone acetylation
proprioception
sensory perception of pain
cerebellum development
ventricular system development
neuron differentiation
negative regulation of histone methylation
neuron projection development
cardiolipin metabolic process
response to estradiol
multicellular organismal response to stress
negative regulation of histone acetylation
social behavior
regulation of gene expression, epigenetic
neuron maturation
negative regulation of neuron apoptotic process
regulation of DNA methylation
negative regulation of transcription, DNA-templated
positive regulation of transcription, DNA-templated
phosphatidylcholine metabolic process
regulation of synaptic plasticity
negative regulation of astrocyte differentiation
catecholamine secretion
neuromuscular process controlling posture
neuromuscular process
positive regulation of synapse assembly
excitatory postsynaptic potential
long-term synaptic potentiation
four-way junction DNA binding
DNA binding
chromatin binding
transcription factor activity, sequence-specific DNA binding
mRNA binding
transcription factor binding
methyl-CpG binding
double-stranded methylated DNA binding
enzyme binding
protein domain specific binding
chromatin DNA binding
siRNA binding
histone deacetylase binding
poly(A) RNA binding
unmethylated CpG binding
protein N-terminus binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0