This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other TEF SIRNA Assays ›
Gene Symbol:
Tef
Gene Name:
thyrotrophic embryonic factor
Gene Aliases:
-
Chromosome Location:
Chr.7: 123033961 - 123058606 on Build Rnor_6.0
UniGene ID:
Rn.59871
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID s131243

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Tef
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_019194.2 NP_062067.2 3 758
XM_006242068.2 XP_006242130.1 3 827
GenBank mRNA BC101895.1 - 3 758 7932583
S58745.1 - 3 581

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_017376, NM_153484, AK196541, BC017689, AK042146, and others.

Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
rhythmic process
RNA polymerase II core promoter proximal region sequence-specific DNA binding
transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
double-stranded DNA binding
protein homodimerization activity
sequence-specific DNA binding
protein heterodimerization activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0