This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other SEPP1 SIRNA Assays ›
Gene Symbol:
Sepp1
Gene Name:
selenoprotein P, plasma, 1
Gene Aliases:
-
Chromosome Location:
Chr.2: 53105912 - 53116198 on Build Rnor_6.0
UniGene ID:
Rn.1451
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID s131237

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Sepp1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001083911.1 NP_001077380.1 5 1080
NM_019192.2 NP_062065.2 5 1008
GenBank mRNA BC059137.1 - 5 1009
BC072539.1 - 5 997 7104195
D25221.1 - 5 989
FQ209666.1 - 1 54
FQ218512.1 - 3 637
FQ218965.1 - 1 350
FQ219620.1 - 1 151
M63574.1 - 968

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

selenium compound metabolic process
brain development
locomotory behavior
post-embryonic development
response to selenium ion
sexual reproduction
growth
selenium binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0