This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other SLC6A18 SIRNA Assays ›
Gene Symbol:
Slc6a18
Gene Name:
solute carrier family 6 (neutral amino acid transporter), member 18
Gene Aliases:
Rosit, Xtrp2
Chromosome Location:
Chr.1: 32221680 - 32235599 on Build Rnor_6.0
UniGene ID:
Rn.9998
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID s131164

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Slc6a18
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_017163.1 NP_058859.1 6, 7 869
XM_006227772.2 XP_006227834.1 6, 7 957
XM_006227774.2 XP_006227836.1 6, 7 957
XM_006227775.2 XP_006227837.1 6, 7 957
XM_008758682.1 XP_008756904.1 5, 6 846
GenBank mRNA U12973.1 - 6, 7 869

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

amino acid transmembrane transport
neurotransmitter transport
amino acid transport
response to osmotic stress
hyperosmotic response
transmembrane transport
neurotransmitter:sodium symporter activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0