This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other TYMS SIRNA Assays ›
Gene Symbol:
Tyms
Gene Name:
thymidylate synthetase
Gene Aliases:
-
Chromosome Location:
Chr.9: 121918875 - 121931564 on Build Rnor_6.0
UniGene ID:
Rn.162284
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID s131047

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Tyms
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_019179.1 NP_062052.1 7 829
XM_008767420.1 XP_008765642.1 7 1318
XM_008767421.1 XP_008765643.1 8 874
GenBank mRNA BC126093.1 - 7 843 8362547
L12138.1 - 7 829

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

pyrimidine nucleobase metabolic process
dTMP biosynthetic process
dTTP biosynthetic process
aging
circadian rhythm
response to toxic substance
response to organic cyclic compound
immortalization of host cell by virus
uracil metabolic process
organ regeneration
methylation
response to progesterone
response to vitamin A
response to cytokine
response to drug
response to ethanol
dUMP metabolic process
tetrahydrofolate metabolic process
response to organophosphorus
developmental growth
cartilage development
response to glucocorticoid
response to folic acid
intestinal epithelial cell maturation
nucleotide binding
mRNA binding
thymidylate synthase activity
folic acid binding
drug binding
protein homodimerization activity
cofactor binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0