This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other POLB SIRNA Assays ›
Gene Symbol:
Polb
Gene Name:
polymerase (DNA directed), beta
Gene Aliases:
-
Chromosome Location:
Chr.16: 74237001 - 74260272 on Build Rnor_6.0
UniGene ID:
Rn.9346
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID s130995

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Polb
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_017141.2 NP_058837.2 6 352
GenBank mRNA BC098668.1 - 6 352 7386894
J02776.1 - 6 340
M13961.1 - 6 330
U38801.1 - 347

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

DNA replication
base-excision repair
base-excision repair, gap-filling
pyrimidine dimer repair
nucleotide-excision repair, DNA gap filling
cellular response to DNA damage stimulus
aging
intrinsic apoptotic signaling pathway in response to DNA damage
response to gamma radiation
response to ethanol
neuron apoptotic process
response to hyperoxia
DNA biosynthetic process
DNA binding
damaged DNA binding
DNA-directed DNA polymerase activity
protein binding
microtubule binding
lyase activity
enzyme binding
metal ion binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-9c87ff496-8j9ll:80/100.66.128.142:80.
git-commit: 146a63324c51a096ecccde4707f5fe4015d9ef66
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36_secrets_rm