This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other VTN SIRNA Assays ›
Gene Symbol:
Vtn
Gene Name:
vitronectin
Gene Aliases:
Aa1018, Vn
Chromosome Location:
Chr.10: 65767960 - 65771040 on Build Rnor_6.0
UniGene ID:
Rn.87493
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID s130825

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Vtn
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_019156.2 NP_062029.2 7 1325
GenBank mRNA AY318963.1 - 1322
BC105821.1 - 7 1325 7371104
FQ209609.1 - 7 1338
FQ209682.1 - 7 1333
FQ209762.1 - 3 499
FQ210146.1 - 5 945
FQ210518.1 - 7 1330
FQ211325.1 - 3 459
FQ212575.1 - 7 1325
FQ218318.1 - 7 1337
FQ218401.1 - 5 897
FQ219297.1 - 7 1339
U44845.1 - 7 1315

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

receptor-mediated endocytosis
immune response
cell-matrix adhesion
positive regulation of cell-substrate adhesion
negative regulation of endopeptidase activity
positive regulation of smooth muscle cell migration
extracellular matrix organization
positive regulation of protein binding
cell adhesion mediated by integrin
endodermal cell differentiation
positive regulation of receptor-mediated endocytosis
positive regulation of peptidyl-tyrosine phosphorylation
smooth muscle cell-matrix adhesion
scavenger receptor activity
integrin binding
polysaccharide binding
extracellular matrix binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-9c87ff496-8j9ll:80/100.66.128.142:80.
git-commit: 146a63324c51a096ecccde4707f5fe4015d9ef66
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36_secrets_rm