This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other VLDLR SIRNA Assays ›
Gene Symbol:
Vldlr
Gene Name:
very low density lipoprotein receptor
Gene Aliases:
-
Chromosome Location:
Chr.1: 245236819 - 245273688 on Build Rnor_6.0
UniGene ID:
Rn.9975
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID s130500

Purification
Size
Availability Made To Order
Catalog # 4390771
Price 368.00
Your Price
Online offer: 309.12
309.1
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Vldlr
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_013155.2 NP_037287.2 8 1344
XM_006231197.2 XP_006231259.1 9 1649
XM_006231198.2 XP_006231260.1 9 1378
XM_006231199.2 XP_006231261.1 8 1252
GenBank mRNA L35767.1 - 8 1344

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

negative regulation of transcription from RNA polymerase II promoter
response to hypoxia
lipid transport
receptor-mediated endocytosis
cell surface receptor signaling pathway
heart development
response to nutrient
cholesterol metabolic process
response to hormone
ventral spinal cord development
cerebral cortex development
response to lipopolysaccharide
cellular response to insulin stimulus
glycoprotein transport
very-low-density lipoprotein particle clearance
reelin-mediated signaling pathway
cellular response to glucose starvation
response to drug
positive regulation of protein kinase activity
positive regulation of protein tyrosine kinase activity
cellular response to lipopolysaccharide
cellular response to interleukin-1
cellular response to hypoxia
positive regulation of dendrite development
glycoprotein binding
calcium ion binding
very-low-density lipoprotein particle receptor activity
protein tyrosine kinase activator activity
apolipoprotein binding
very-low-density lipoprotein particle binding
glycoprotein transporter activity
reelin receptor activity
calcium-dependent protein binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-7c5f86c6b-2xgzq:80/100.66.128.142:80.
git-commit: c124ca8d7fcef91bd439783ab4009544f5b84403
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0