This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other ITPR3 SIRNA Assays ›
Gene Symbol:
Itpr3
Gene Name:
inositol 1,4,5-trisphosphate receptor, type 3
Gene Aliases:
IP3R3, IP3R3X
Chromosome Location:
Chr.20: 5645942 - 5711707 on Build Rnor_6.0
UniGene ID:
Rn.11242
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID s130454

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Itpr3
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_013138.1 NP_037270.1 48 6534
XM_008772710.1 XP_008770932.1 49 6598
GenBank mRNA GQ233031.1 - 48 6400
L06096.1 - 48 6534

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_080553, AK028491, BC023776, AB182289.

Gene Ontology Categories:

Function(s) Process(es)

calcium ion transport
G-protein coupled receptor signaling pathway
positive regulation of cytosolic calcium ion concentration
memory
inositol phosphate-mediated signaling
sensory perception of bitter taste
sensory perception of sweet taste
sensory perception of umami taste
protein homooligomerization
protein heterooligomerization
response to calcium ion
long-term synaptic potentiation
calcium ion transport into cytosol
calcium ion transmembrane transport
cellular response to cAMP
inositol hexakisphosphate binding
inositol 1,4,5-trisphosphate-sensitive calcium-release channel activity
calcium ion binding
calcium ion transmembrane transporter activity
calcium-release channel activity
phosphatidylinositol binding
inositol 1,3,4,5 tetrakisphosphate binding
inositol 1,4,5 trisphosphate binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0