This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other BMP3 SIRNA Assays ›
Gene Symbol:
Bmp3
Gene Name:
bone morphogenetic protein 3
Gene Aliases:
PBMP3
Chromosome Location:
Chr.14: 12362912 - 12387572 on Build Rnor_6.0
UniGene ID:
Rn.208979
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID s130419

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Bmp3
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_017105.1 NP_058801.1 2 1593
GenBank mRNA D29768.1 - 1 142
D63860.1 - 2 1593

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

ossification
positive regulation of pathway-restricted SMAD protein phosphorylation
growth
regulation of apoptotic process
regulation of MAPK cascade
positive regulation of transcription from RNA polymerase II promoter
cell development
cartilage development
SMAD protein signal transduction
cytokine activity
transforming growth factor beta receptor binding
growth factor activity
BMP receptor binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0