This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other IL1R1 SIRNA Assays ›
Gene Symbol:
Il1r1
Gene Name:
interleukin 1 receptor, type I
Gene Aliases:
-
Chromosome Location:
Chr.9: 46962292 - 47038139 on Build Rnor_6.0
UniGene ID:
Rn.9758
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID s130407

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Il1r1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_013123.3 NP_037255.3 8 1026
XM_006244746.2 XP_006244808.1 9 3135
XM_006244747.2 XP_006244809.1 9 1392
XM_006244748.2 XP_006244810.1 8 1296
XM_006244749.2 XP_006244811.1 10 1215
XM_006244750.2 XP_006244812.1 10 3207
XM_006244751.2 XP_006244813.1 9 1126
XM_006244752.2 XP_006244814.1 9 1368
XM_006244753.2 XP_006244815.1 9 2487
XM_006244754.2 XP_006244816.1 8 3039
XM_008766993.1 XP_008765215.1 9 1143
GenBank mRNA AB079118.1 - 6 720
AB112436.1 - 10 1160
AB112437.1 - 8 1026
AB112438.1 - 9 1098
M95578.1 - 9 1347

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

response to radiation
heat acclimation
cytokine-mediated signaling pathway
ovulation
regulation of inflammatory response
interleukin-1-mediated signaling pathway
response to interleukin-1
response to epidermal growth factor
cellular response to glucose stimulus
response to transforming growth factor beta
response to nitric oxide
protease binding
signal transducer activity
interleukin-1 receptor activity
interleukin-1, Type I, activating receptor activity
platelet-derived growth factor receptor binding
protein binding
ionotropic glutamate receptor binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0