This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other TSHB SIRNA Assays ›
Gene Symbol:
Tshb
Gene Name:
thyroid stimulating hormone, beta
Gene Aliases:
-
Chromosome Location:
Chr.2: 205206457 - 205214812 on Build Rnor_6.0
UniGene ID:
Rn.9966
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID s130382

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Tshb
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_013116.2 NP_037248.2 2, 3 226
XM_008761373.1 XP_008759595.1 2, 3 2357
GenBank mRNA BC058488.1 - 2, 3 186 6921292
CB315383.1 - 2, 3 185
D00578.1 - 1, 2 39
M10902.1 - 2, 3 181
X01454.1 - 2, 3 224

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_001165940, NM_009432, NM_001165939, J00644, M54943, and others.

Gene Ontology Categories:

Function(s) Process(es)

response to vitamin A
response to estrogen
response to calcium ion
hormone activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0