This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other SDC2 SIRNA Assays ›
Gene Symbol:
Sdc2
Gene Name:
syndecan 2
Gene Aliases:
HSPG
Chromosome Location:
Chr.7: 71572731 - 71686139 on Build Rnor_6.0
UniGene ID:
Rn.11127
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID s130272

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Sdc2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_013082.3 NP_037214.1 3 599
XM_006241518.2 XP_006241580.1 3 811
GenBank mRNA BC070890.1 - 3 389 7098631
FQ218442.1 - 3 332
FQ219283.1 - 3 633
FQ219510.1 - 3 631
FQ222974.1 - 3 681
M81687.1 - 3 599

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_008304, AK011042, U00674, BC047144, AK216657, and others.

Gene Ontology Categories:

Function(s) Process(es)

response to hypoxia
response to caffeine
wound healing
dendrite morphogenesis
regulation of dendrite morphogenesis
protein binding
PDZ domain binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0