This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other YWHAH SIRNA Assays ›
Gene Symbol:
Ywhah
Gene Name:
tyrosine 3-monooxygenase/tryptophan 5-monooxygenase activation protein, eta
Gene Aliases:
14-3-3e
Chromosome Location:
Chr.14: 83053041 - 83062424 on Build Rnor_6.0
UniGene ID:
Rn.3324
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID s130170

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Ywhah
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_013052.1 NP_037184.1 2 363
GenBank mRNA BC081825.1 - 2 388 7192904
D17445.1 - 2 363

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_011738, BC008187, BC061497, AK187410, D87661, and others.

Gene Ontology Categories:

Function(s) Process(es)

regulation of sodium ion transport
glucocorticoid catabolic process
intracellular protein transport
biological_process
substantia nigra development
glucocorticoid receptor signaling pathway
positive regulation of transcription, DNA-templated
negative regulation of dendrite morphogenesis
membrane depolarization during action potential
regulation of sodium ion transmembrane transporter activity
molecular_function
actin binding
protein binding
sodium channel regulator activity
enzyme binding
protein domain specific binding
glucocorticoid receptor binding
ion channel binding
protein heterodimerization activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0