siRNA Details
Gene Symbol: Ttpa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013048.2 | NP_037180.1 | 3 | 465 | ||
XM_008763588.1 | XP_008761810.1 | 5 | 472 | |||
XM_008763589.1 | XP_008761811.1 | 4 | 415 |

Gene Symbol: Ttpa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013048.2 | NP_037180.1 | 3 | 465 | ||
XM_008763588.1 | XP_008761810.1 | 5 | 472 | |||
XM_008763589.1 | XP_008761811.1 | 4 | 415 |