This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other SHOX2 SIRNA Assays ›
Gene Symbol:
Shox2
Gene Name:
short stature homeobox 2
Gene Aliases:
-
Chromosome Location:
Chr.2: 164118175 - 164126783 on Build Rnor_6.0
UniGene ID:
Rn.11258
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID s130084

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Shox2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_013028.1 NP_037160.1 4, 5 445
GenBank mRNA AJ002259.1 - 4, 5 445

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Human: NM_003030, NM_006884, BC008829, AJ002367.
This siRNA also targets the following transcripts in Mouse: NM_013665, AK045937, U66918.

Gene Ontology Categories:

Function(s) Process(es)

negative regulation of transcription from RNA polymerase II promoter
osteoblast differentiation
positive regulation of mesenchymal cell proliferation
chondrocyte differentiation
chondrocyte development
heart valve development
cardiac atrium morphogenesis
regulation of transcription, DNA-templated
multicellular organism development
embryonic limb morphogenesis
regulation of chondrocyte differentiation
embryonic forelimb morphogenesis
positive regulation of smoothened signaling pathway
positive regulation of transcription from RNA polymerase II promoter
embryonic digestive tract morphogenesis
embryonic morphogenesis
positive regulation of skeletal muscle fiber development
positive regulation of axonogenesis
embryonic skeletal joint morphogenesis
cartilage development involved in endochondral bone morphogenesis
muscle tissue morphogenesis
regulation of branching morphogenesis of a nerve
sequence-specific DNA binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0