This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other SELE SIRNA Assays ›
Gene Symbol:
Sele
Gene Name:
selectin E
Gene Aliases:
-
Chromosome Location:
Chr.13: 82355229 - 82365323 on Build Rnor_6.0
UniGene ID:
Rn.10359
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID s130076

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Sele
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_138879.1 NP_620234.1 2 226
XM_008769608.1 XP_008767830.1 3 608
GenBank mRNA L25527.1 - 2 226
X69981.1 - 2 278

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_011345, AK153623, BC120795, M87862, BC119083.

Gene Ontology Categories:

Function(s) Process(es)

positive regulation of receptor internalization
positive regulation of leukocyte migration
inflammatory response
cell adhesion
heterophilic cell-cell adhesion via plasma membrane cell adhesion molecules
leukocyte cell-cell adhesion
activation of phospholipase C activity
actin filament-based process
response to cytokine
leukocyte migration
leukocyte tethering or rolling
response to interleukin-1
molecular_function
transmembrane signaling receptor activity
carbohydrate binding
sialic acid binding
phospholipase binding
oligosaccharide binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0