This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other SAG SIRNA Assays ›
Gene Symbol:
Sag
Gene Name:
S-antigen; retina and pineal gland (arrestin)
Gene Aliases:
SAGMR, SANTI
Chromosome Location:
Chr.9: 94926838 - 94972162 on Build Rnor_6.0
UniGene ID:
Rn.9856
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID s130066

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Sag
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_013023.2 NP_037155.2 16 1285
XM_008767196.1 XP_008765418.1 16 1548
GenBank mRNA M60737.1 - 15 1176
X15353.1 - 15 1176
X51781.1 - 16 1285

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

signal transduction
visual perception
biological_process
opsin binding
molecular_function
spectrin binding
phosphoprotein binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0