This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other LGALS4 SIRNA Assays ›
Gene Symbol:
Lgals4
Gene Name:
lectin, galactoside-binding, soluble, 4
Gene Aliases:
L-36, L36LBI
Chromosome Location:
Chr.1: 87019808 - 87027720 on Build Rnor_6.0
UniGene ID:
Rn.9656
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID s129891

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Lgals4
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_012975.1 NP_037107.1 5 505
XM_008759113.1 XP_008757335.1 5 672
GenBank mRNA M73553.1 - 5 505

Back To Top

More Information


Gene Ontology Categories:

Process(es)

carbohydrate binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-9c87ff496-8j9ll:80/100.66.128.142:80.
git-commit: 146a63324c51a096ecccde4707f5fe4015d9ef66
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36_secrets_rm