This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other IL1RAP SIRNA Assays ›
Gene Symbol:
Il1rap
Gene Name:
interleukin 1 receptor accessory protein
Gene Aliases:
IL-1RAcp, Il1racpb
Chromosome Location:
Chr.11: 77463242 - 77593207 on Build Rnor_6.0
UniGene ID:
Rn.10511
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID s129866

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Il1rap
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001167840.1 NP_001161312.1 10 1360
NM_012968.1 NP_037100.1 9 1279
XM_006248504.2 XP_006248566.1 9 1348
GenBank mRNA GU123169.1 - 10 1360
U48592.1 - 9 1279

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

cytokine-mediated signaling pathway
interleukin-2 biosynthetic process
signal transducer activity
interleukin-1 receptor activity
interleukin-1 receptor binding
interleukin-1 binding
protein binding, bridging

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0