This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other GDNF SIRNA Assays ›
Gene Symbol:
Gdnf
Gene Name:
glial cell derived neurotrophic factor
Gene Aliases:
gndf
Chromosome Location:
Chr.2: 56887546 - 56912964 on Build Rnor_6.0
UniGene ID:
Rn.53970
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID s129837

Purification
Size
Availability Made To Order
Catalog # 4390771
Price 368.00
Your Price
Online offer: 309.12
309.1
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Gdnf
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_019139.1 NP_062012.1 3 546
XM_006232010.2 XP_006232072.1 3 987
XM_006232011.2 XP_006232073.1 3 645
XM_008760756.1 XP_008758978.1 2 1824
XM_008760757.1 XP_008758979.1 2 1746
GenBank mRNA AF497634.1 - 2 474
EU068467.1 - 2 434
EU068468.1 - 2 434
EU068469.1 - 2 512
EU068470.1 - 2 497
EU068471.1 - 2 497
EU068472.1 - 2 497
L15305.1 - 3 546
S75583.1 - 2 497

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

metanephros development
ureteric bud development
branching involved in ureteric bud morphogenesis
neural crest cell migration
organ induction
postsynaptic membrane organization
mesenchymal to epithelial transition involved in metanephros morphogenesis
nervous system development
peripheral nervous system development
cell proliferation
positive regulation of cell proliferation
male gonad development
response to wounding
regulation of gene expression
postganglionic parasympathetic fiber development
neuron differentiation
peristalsis
midbrain development
neuron projection development
positive regulation of monooxygenase activity
negative regulation of neuron apoptotic process
positive regulation of cell differentiation
positive regulation of transcription from RNA polymerase II promoter
mRNA stabilization
enteric nervous system development
sympathetic nervous system development
regulation of dopamine uptake involved in synaptic transmission
retina development in camera-type eye
ureteric bud formation
regulation of morphogenesis of a branching structure
cellular response to dexamethasone stimulus
regulation of ureteric bud formation
positive regulation of ureteric bud formation
positive regulation of mesenchymal to epithelial transition involved in metanephros morphogenesis
positive regulation of branching involved in ureteric bud morphogenesis
negative regulation of extrinsic apoptotic signaling pathway in absence of ligand
growth factor activity
glial cell-derived neurotrophic factor receptor binding
protein homodimerization activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0