siRNA Details
Gene Symbol: F2r | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012950.2 | NP_037082.2 | 2 | 171 | ||
GenBank mRNA | BC085759.1 | - | 2 | 171 | 7106133 | |
M81642.1 | - | 2 | 179 |

Gene Symbol: F2r | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012950.2 | NP_037082.2 | 2 | 171 | ||
GenBank mRNA | BC085759.1 | - | 2 | 171 | 7106133 | |
M81642.1 | - | 2 | 179 |