This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other DPYSL3 SIRNA Assays ›
Gene Symbol:
Dpysl3
Gene Name:
dihydropyrimidinase-like 3
Gene Aliases:
Crmp4, TUC-4b
Chromosome Location:
Chr.18: 37716433 - 37819347 on Build Rnor_6.0
UniGene ID:
Rn.93365
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID s129746

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Dpysl3
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_012934.1 NP_037066.1 9 938
XM_006254670.2 XP_006254732.1 9 1518
XM_006254671.2 XP_006254733.1 9 1149
GenBank mRNA AF389425.1 - 9 938
AF398465.1 - 9 1207
U52104.1 - 9 1154

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

pyrimidine nucleobase catabolic process
nervous system development
positive regulation of neuron projection development
negative regulation of neuron projection development
negative regulation of cell migration
neuron development
response to axon injury
actin filament bundle assembly
protein homooligomerization
positive regulation of filopodium assembly
actin crosslink formation
cellular response to cytokine stimulus
hydrolase activity, acting on carbon-nitrogen (but not peptide) bonds, in cyclic amides
SH3 domain binding
chondroitin sulfate binding
phosphoprotein binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0