This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other BFSP1 SIRNA Assays ›
Gene Symbol:
Bfsp1
Gene Name:
beaded filament structural protein 1, filensin
Gene Aliases:
115-kDa, CP94, CP95, CP97
Chromosome Location:
Chr.3: 137935345 - 137984999 on Build Rnor_6.0
UniGene ID:
Rn.9785
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID s129681

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Bfsp1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_031555.1 NP_113743.1 8 1967
XM_006235117.2 XP_006235179.1 8 1628
GenBank mRNA AB003104.1 - 8 1968

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

cell maturation
lens fiber cell development
structural constituent of eye lens

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-7c5f86c6b-2xgzq:80/100.66.128.142:80.
git-commit: c124ca8d7fcef91bd439783ab4009544f5b84403
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0