This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other ARRB1 SIRNA Assays ›
Gene Symbol:
Arrb1
Gene Name:
arrestin, beta 1
Gene Aliases:
BARRES
Chromosome Location:
Chr.1: 164502354 - 164573947 on Build Rnor_6.0
UniGene ID:
Rn.34876
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID s129662

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Arrb1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_012910.2 NP_037042.1 14 1076
XM_006229734.2 XP_006229796.1 14 1702
XM_006229735.2 XP_006229797.1 13 1677
XM_006229736.2 XP_006229798.1 14 1671
XM_006229737.2 XP_006229799.1 13 1646
XM_006229738.2 XP_006229800.1 14 1703
XM_006229739.2 XP_006229801.1 14 1818
XM_006229740.2 XP_006229802.1 14 1204
XM_006229741.2 XP_006229803.1 14 1217
GenBank mRNA M91589.1 - 14 1079

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

activation of MAPK activity
negative regulation of protein phosphorylation
positive regulation of protein phosphorylation
G-protein coupled receptor internalization
desensitization of G-protein coupled receptor protein signaling pathway by arrestin
positive regulation of receptor internalization
transcription from RNA polymerase II promoter
endocytosis
receptor-mediated endocytosis
G-protein coupled receptor signaling pathway
adenylate cyclase-modulating G-protein coupled receptor signaling pathway
phototransduction
regulation of G-protein coupled receptor protein signaling pathway
protein transport
protein ubiquitination
sensory perception of pain
regulation of cAMP biosynthetic process
regulation of cAMP catabolic process
negative regulation of protein ubiquitination
positive regulation of protein ubiquitination
negative regulation of NF-kappaB transcription factor activity
positive regulation of protein binding
negative regulation of interleukin-6 production
negative regulation of interleukin-8 production
positive regulation of peptidyl-serine phosphorylation
negative regulation of GTPase activity
positive regulation of smooth muscle cell apoptotic process
positive regulation of Rho protein signal transduction
positive regulation of histone acetylation
positive regulation of insulin secretion involved in cellular response to glucose stimulus
follicle-stimulating hormone signaling pathway
stress fiber assembly
negative regulation of cysteine-type endopeptidase activity involved in apoptotic process
proteasome-mediated ubiquitin-dependent protein catabolic process
positive regulation of cysteine-type endopeptidase activity involved in apoptotic process
positive regulation of GTPase activity
positive regulation of transcription from RNA polymerase II promoter
sensory perception of touch
negative regulation of ERK1 and ERK2 cascade
positive regulation of ERK1 and ERK2 cascade
positive regulation of histone H4 acetylation
G-protein coupled receptor binding
histone acetyltransferase activity
GTPase activator activity
insulin-like growth factor receptor binding
protein binding
transcription factor binding
clathrin binding
estrogen receptor binding
mitogen-activated protein kinase kinase binding
ubiquitin protein ligase binding
alpha-1A adrenergic receptor binding
alpha-1B adrenergic receptor binding
angiotensin receptor binding
follicle-stimulating hormone receptor binding
V2 vasopressin receptor binding
AP-2 adaptor complex binding
clathrin adaptor activity
cysteine-type endopeptidase inhibitor activity involved in apoptotic process
transcription regulatory region DNA binding
ion channel binding
protein phosphorylated amino acid binding
phosphoprotein binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-7c5f86c6b-2xgzq:80/100.66.128.142:80.
git-commit: c124ca8d7fcef91bd439783ab4009544f5b84403
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0