This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other SLC30A2 SIRNA Assays ›
Gene Symbol:
Slc30a2
Gene Name:
solute carrier family 30 (zinc transporter), member 2
Gene Aliases:
ZnT-2, Znt2
Chromosome Location:
Chr.5: 152559577 - 152571800 on Build Rnor_6.0
UniGene ID:
Rn.11135
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID s129594

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Slc30a2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001083122.1 NP_001076591.1 2, 3 329
NM_012890.2 NP_037022.1 1, 2 268
GenBank mRNA BC061997.1 - 1, 2 271
BC168237.1 - 2, 3 322 7938890
CB778290.1 - 2, 3 332
U50927.1 - 1, 2 268

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

zinc II ion transport
positive regulation of sequestering of zinc ion
zinc II ion transmembrane transport
cation transmembrane transport
zinc ion transmembrane transporter activity
cation transmembrane transporter activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0