This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other NFYB SIRNA Assays ›
Gene Symbol:
Nfyb
Gene Name:
nuclear transcription factor-Y beta
Gene Aliases:
CBF-B, CBFB, Nfya
Chromosome Location:
Chr.7: 27081667 - 27097481 on Build Rnor_6.0
UniGene ID:
Rn.90077
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID s129520

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Nfyb
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_031553.2 NP_113741.1 4 334
XM_006241196.2 XP_006241258.1 4 352
GenBank mRNA BC089791.1 - 4 334 7380395
M55045.1 - 3 203

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Human: NM_006166, AK291934, BG776259, BC005317, BC005316, and others.
This siRNA also targets the following transcripts in Mouse: NM_010914, AK154749, BB858171, AK155749, AK146168, and others.

Gene Ontology Categories:

Function(s) Process(es)

regulation of transcription, DNA-templated
transcription from RNA polymerase II promoter
positive regulation of transcription, DNA-templated
DNA binding
transcription factor activity, sequence-specific DNA binding
protein complex binding
sequence-specific DNA binding
transcription regulatory region DNA binding
protein heterodimerization activity
repressing transcription factor binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0