This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other MAT1A SIRNA Assays ›
Gene Symbol:
Mat1a
Gene Name:
methionine adenosyltransferase I, alpha
Gene Aliases:
AdoMet, SADE, SAS
Chromosome Location:
Chr.16: 18690649 - 18709135 on Build Rnor_6.0
UniGene ID:
Rn.10418
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID s129504

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Mat1a
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_012860.2 NP_036992.2 6 781
GenBank mRNA BC089770.1 - 6 781 7368255
X15734.1 - 6 688
X60822.1 - 6 867

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Human: NM_000429, D49357, BC018359, AK290820, X69078.
This siRNA also targets the following transcripts in Mouse: NM_133653, BC011211, L13622, AK149542, AK153832.

Gene Ontology Categories:

Function(s) Process(es)

S-adenosylmethionine biosynthetic process
one-carbon metabolic process
protein homooligomerization
protein tetramerization
protein homotetramerization
magnesium ion binding
methionine adenosyltransferase activity
ATP binding
amino acid binding
identical protein binding
protein homodimerization activity
ADP binding
protein dimerization activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0