This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other CYCT SIRNA Assays ›
Gene Symbol:
Cyct Pde11a
Gene Name:
cytochrome c, testis
phosphodiesterase 11A
Gene Aliases:
CYCTA
-
Chromosome Location:
Chr.3: 63204780 - 63207962 on Build Rnor_6.0
Chromosome Location:
Chr.3: 62818502 - 63211843 on Build Rnor_6.0
UniGene ID:
Rn.88630
Rn.88630
Species:
Rat
Species Specific ID (Flybase ID):
-
-
siRNA ID s129441

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Cyct
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_012840.3 NP_036972.1 369
GenBank mRNA M20623.1 - 1 199
Gene Symbol: Pde11a
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001127480.1 NP_001120952.1 3 371
GenBank mRNA AB059361.1 - 3 371

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_009989, AK005582, AK018851, BC049608, X55771, and others.

Gene Ontology Categories:

Function(s) Process(es)

apoptotic process
signal transduction
metabolic process
oxidation-reduction process
cAMP catabolic process
cGMP catabolic process
3',5'-cyclic-nucleotide phosphodiesterase activity
iron ion binding
electron carrier activity
heme binding
metal ion binding
cyclic-nucleotide phosphodiesterase activity
cGMP-stimulated cyclic-nucleotide phosphodiesterase activity
cAMP binding
cGMP binding
3',5'-cyclic-GMP phosphodiesterase activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-67c6c7f55f-2g6mz:80/100.66.128.142:80.
git-commit: 92b42c745e7842b39e6d37af9c4064bd81a92e4d
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header