This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other SMO SIRNA Assays ›
Gene Symbol:
Smo
Gene Name:
smoothened, frizzled class receptor
Gene Aliases:
Smoh
Chromosome Location:
Chr.4: 57019941 - 57041779 on Build Rnor_6.0
UniGene ID:
Rn.44429
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID s129340

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Smo
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_012807.1 NP_036939.1 5 967
GenBank mRNA U84402.1 - 5 967

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Human: NM_005631, AF071494, BC009989, AF120103, U84401.

Gene Ontology Categories:

Function(s) Process(es)

negative regulation of transcription from RNA polymerase II promoter
ossification
vasculogenesis
osteoblast differentiation
in utero embryonic development
cell fate specification
neural crest cell migration
heart looping
positive regulation of neuroblast proliferation
positive regulation of mesenchymal cell proliferation
heart morphogenesis
determination of left/right asymmetry in lateral mesoderm
type B pancreatic cell development
G-protein coupled receptor signaling pathway
smoothened signaling pathway
positive regulation of hh target transcription factor activity
spermatogenesis
determination of left/right symmetry
ventral midline determination
pattern specification process
central nervous system development
midgut development
positive regulation of cell proliferation
response to wounding
anterior/posterior pattern specification
dorsal/ventral pattern formation
response to organic substance
regulation of gene expression
positive regulation of gene expression
negative regulation of gene expression
facial nerve development
cerebellar cortex morphogenesis
thalamus development
dorsal/ventral neural tube patterning
smoothened signaling pathway involved in ventral spinal cord patterning
smoothened signaling pathway involved in regulation of cerebellar granule cell precursor cell proliferation
central nervous system neuron differentiation
cerebral cortex development
negative regulation of epithelial cell differentiation
hair follicle morphogenesis
neuron projection regeneration
osteoblast proliferation
protein localization to nucleus
multicellular organism growth
non-canonical Wnt signaling pathway
positive regulation of protein import into nucleus
odontogenesis of dentin-containing tooth
negative regulation of apoptotic process
negative regulation of DNA binding
positive regulation of smoothened signaling pathway
positive regulation of transcription from RNA polymerase II promoter
astrocyte activation
cell development
digestive tract development
embryonic organ development
developmental growth
skeletal muscle fiber development
axon extension involved in axon guidance
forebrain morphogenesis
homeostasis of number of cells within a tissue
positive regulation of epithelial cell proliferation
protein stabilization
myoblast migration
negative regulation of hair follicle development
canonical Wnt signaling pathway
detection of cell density by contact stimulus involved in contact inhibition
atrial septum morphogenesis
mammary gland epithelial cell differentiation
epithelial-mesenchymal cell signaling
somite development
pancreas morphogenesis
left/right axis specification
cellular response to cholesterol
renal system development
mesenchymal to epithelial transition involved in metanephric renal vesicle formation
positive regulation of branching involved in ureteric bud morphogenesis
regulation of stem cell population maintenance
regulation of heart morphogenesis
molecular_function
transmembrane signaling receptor activity
G-protein coupled receptor activity
patched binding
drug binding
Wnt-protein binding
Wnt-activated receptor activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0