This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other GPC3 SIRNA Assays ›
Gene Symbol:
Gpc3
Gene Name:
glypican 3
Gene Aliases:
OCI-5
Chromosome Location:
Chr.X: 139579268 - 139947093 on Build Rnor_6.0
UniGene ID:
Rn.9717
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID s129231

Purification
Size
Availability Made To Order
Catalog # 4390771
Price 368.00
Your Price
Online offer: 309.12
309.1
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Gpc3
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_012774.1 NP_036906.1 6 1432
GenBank mRNA BC085756.1 - 6 1447 7122343
M22400.1 - 6 1432

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

branching involved in ureteric bud morphogenesis
kidney development
biological_process
negative regulation of cell proliferation
organ morphogenesis
anterior/posterior axis specification
body morphogenesis
negative regulation of peptidase activity
bone mineralization
osteoclast differentiation
lung development
positive regulation of BMP signaling pathway
embryonic hindlimb morphogenesis
regulation of growth
positive regulation of protein catabolic process
positive regulation of endocytosis
negative regulation of smoothened signaling pathway
positive regulation of smoothened signaling pathway
negative regulation of growth
positive regulation of glucose import
negative regulation of epithelial cell proliferation
coronary vasculature development
cell proliferation involved in kidney development
mesenchymal cell proliferation involved in ureteric bud development
mesonephric duct morphogenesis
cell proliferation involved in metanephros development
negative regulation of canonical Wnt signaling pathway
positive regulation of Wnt signaling pathway, planar cell polarity pathway
molecular_function
heparan sulfate proteoglycan binding
peptidyl-dipeptidase inhibitor activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-7db4fb6694-gx648:80/100.66.128.142:80.
git-commit: 672a5a8c50d6b616e949be631a941c86748ade1b
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header