This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other SLC25A5 SIRNA Assays ›
Gene Symbol:
Slc25a5 LOC689073
Gene Name:
solute carrier family 25 (mitochondrial carrier; adenine nucleotide translocator), member 5
similar to solute carrier family 25, member 5
Gene Aliases:
Ant2
-
Chromosome Location:
Chr.X: 123404570 - 123407637 on Build Rnor_6.0
Chromosome Location:
Chr.1: 219748353 - 219749704 on Build Rnor_6.0
UniGene ID:
Rn.102263
-
Species:
Rat
Species Specific ID (Flybase ID):
-
-
siRNA ID s129140

Purification
Size
Availability Made To Order
Catalog # 4390771
Price 368.00
Your Price
Online offer: 309.12
309.1
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Slc25a5
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_057102.1 NP_476443.1 4 839
GenBank mRNA BC059108.1 - 4 888 6922359
D12771.1 - 4 839
FQ213290.1 - 4 861
FQ213647.1 - 4 861
FQ213855.1 - 4 879
FQ214507.1 - 4 816
FQ219677.1 - 4 861
FQ220249.1 - 4 863
FQ220272.1 - 4 864
FQ220435.1 - 4 883
FQ229325.1 - 4 864
FQ229438.1 - 4 864
Gene Symbol: LOC689073
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_008760193.1 XP_008758415.1 2 872

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

chromosome segregation
positive regulation of cell proliferation
transmembrane transport
negative regulation of mitochondrial outer membrane permeabilization involved in apoptotic signaling pathway
transporter activity
protein binding
poly(A) RNA binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0