This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other SLC25A5 SIRNA Assays ›
Gene Symbol:
Slc25a5
Gene Name:
solute carrier family 25 (mitochondrial carrier; adenine nucleotide translocator), member 5
Gene Aliases:
Ant2
Chromosome Location:
Chr.X: 123404570 - 123407637 on Build Rnor_6.0
UniGene ID:
Rn.102263
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID s129139

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Slc25a5
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_057102.1 NP_476443.1 3, 4 798
GenBank mRNA BC059108.1 - 3, 4 847 6922359
D12771.1 - 3, 4 798
FQ213290.1 - 3, 4 820
FQ213647.1 - 3, 4 820
FQ213855.1 - 3, 4 838
FQ214507.1 - 3, 4 775
FQ219677.1 - 3, 4 820
FQ220249.1 - 3, 4 822
FQ220272.1 - 3, 4 823
FQ220435.1 - 3, 4 842
FQ229325.1 - 3, 4 823
FQ229438.1 - 3, 4 823

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

chromosome segregation
positive regulation of cell proliferation
transmembrane transport
negative regulation of mitochondrial outer membrane permeabilization involved in apoptotic signaling pathway
transporter activity
protein binding
poly(A) RNA binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-67c6c7f55f-2g6mz:80/100.66.128.142:80.
git-commit: 92b42c745e7842b39e6d37af9c4064bd81a92e4d
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header