This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other FGFR4 SIRNA Assays ›
Gene Symbol:
Fgfr4
Gene Name:
fibroblast growth factor receptor 4
Gene Aliases:
-
Chromosome Location:
Chr.17: 9990072 - 10004335 on Build Rnor_6.0
UniGene ID:
Rn.24104
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID s129018

Purification
Size
Availability Made To Order
Catalog # 4390771
Price 368.00
Your Price
Online offer: 309.12
309.1
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Fgfr4
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001109904.1 NP_001103374.1 9 1277
XM_006253606.2 XP_006253668.1 9 1294
XM_008771485.1 XP_008769707.1 9 1288
GenBank mRNA BC100260.1 - 8 1369 7100540
M91599.1 - 6 743

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

organ induction
positive regulation of cell proliferation
fibroblast growth factor receptor signaling pathway
positive regulation of gene expression
regulation of extracellular matrix disassembly
cell migration
peptidyl-tyrosine phosphorylation
regulation of lipid metabolic process
lung development
glucose homeostasis
positive regulation of catalytic activity
positive regulation of proteolysis
protein autophosphorylation
phosphate ion homeostasis
alveolar secondary septum development
positive regulation of ERK1 and ERK2 cascade
regulation of bile acid biosynthetic process
response to bile acid
regulation of cholesterol homeostasis
positive regulation of DNA biosynthetic process
fibroblast growth factor-activated receptor activity
ATP binding
heparin binding
fibroblast growth factor binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0