This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other TH SIRNA Assays ›
Gene Symbol:
Th
Gene Name:
tyrosine hydroxylase
Gene Aliases:
The
Chromosome Location:
Chr.1: 216073034 - 216080287 on Build Rnor_6.0
UniGene ID:
Rn.11082
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID s128949

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Th
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_012740.3 NP_036872.1 10, 11 1125
GenBank mRNA M10244.1 - 10, 11 1125

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_009377, M69200, AK139568.

Gene Ontology Categories:

Function(s) Process(es)

response to hypoxia
synaptic transmission, dopaminergic
response to amphetamine
dopamine biosynthetic process from tyrosine
fatty acid metabolic process
sphingolipid metabolic process
heart development
visual perception
sensory perception of sound
learning
memory
mating behavior
locomotory behavior
regulation of heart contraction
response to water deprivation
response to light stimulus
response to herbicide
response to salt stress
organ morphogenesis
response to metal ion
response to zinc ion
multicellular organism aging
response to organic cyclic compound
response to activity
aminergic neurotransmitter loading into synaptic vesicle
glycoside metabolic process
response to insecticide
phthalate metabolic process
cerebral cortex development
response to nutrient levels
response to estradiol
response to lipopolysaccharide
isoquinoline alkaloid metabolic process
response to nicotine
social behavior
cellular response to drug
response to isolation stress
response to immobilization stress
neurotransmitter biosynthetic process
terpene metabolic process
dopamine biosynthetic process
epinephrine biosynthetic process
norepinephrine biosynthetic process
catecholamine biosynthetic process
eye photoreceptor cell development
response to drug
circadian sleep/wake cycle
eating behavior
response to peptide hormone
response to ethanol
response to ether
response to pyrethroid
response to steroid hormone
embryonic camera-type eye morphogenesis
cognition
protein homotetramerization
response to corticosterone
response to electrical stimulus
phytoalexin metabolic process
oxidation-reduction process
response to growth factor
cellular response to manganese ion
cellular response to alkaloid
cellular response to nicotine
cellular response to glucose stimulus
cellular response to growth factor stimulus
monooxygenase activity
tyrosine 3-monooxygenase activity
protein binding
ferrous iron binding
ferric iron binding
amino acid binding
oxygen binding
enzyme binding
protein domain specific binding
tetrahydrobiopterin binding
dopamine binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0