This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other HNF1A SIRNA Assays ›
Gene Symbol:
Hnf1a
Gene Name:
HNF1 homeobox A
Gene Aliases:
HNF1, LF-B1, Lfb1, Tcf1
Chromosome Location:
Chr.12: 47407811 - 47433342 on Build Rnor_6.0
UniGene ID:
Rn.9660
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID s128490

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Hnf1a
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_012669.1 NP_036801.1 10 1994
GenBank mRNA J03170.1 - 10 1994
X53297.1 - 9 1352
X54423.1 - 10 1844

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

negative regulation of transcription from RNA polymerase II promoter
blastocyst development
liver development
placenta development
chromatin remodeling
regulation of transcription, DNA-templated
regulation of transcription from RNA polymerase II promoter
transcription from RNA polymerase II promoter
fatty acid biosynthetic process
bile acid biosynthetic process
heme biosynthetic process
response to oxidative stress
protein localization
cholesterol metabolic process
response to glucose
bile acid and bile salt transport
fatty acid transport
insulin secretion
regulation of Wnt signaling pathway
embryonic limb morphogenesis
endocrine pancreas development
regulation of pronephros size
renal glucose absorption
glucose homeostasis
reverse cholesterol transport
bone resorption
positive regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
glucose import
regulation of hormone secretion
paraxial mesoderm formation
reproductive structure development
regulation of insulin secretion
positive regulation of transcription initiation from RNA polymerase II promoter
SMAD protein signal transduction
RNA polymerase II core promoter sequence-specific DNA binding
transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
transcription cofactor binding
transcriptional activator activity, RNA polymerase II transcription regulatory region sequence-specific binding
DNA binding
double-stranded DNA binding
transcription factor activity, sequence-specific DNA binding
protein binding
transcription factor binding
protein homodimerization activity
sequence-specific DNA binding
transcription regulatory region DNA binding
protein heterodimerization activity
protein dimerization activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0