This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other VAMP2 SIRNA Assays ›
Gene Symbol:
Vamp2
Gene Name:
vesicle-associated membrane protein 2
Gene Aliases:
RATVAMPB, RATVAMPIR, SYB, Syb2
Chromosome Location:
Chr.10: 55675119 - 55679405 on Build Rnor_6.0
UniGene ID:
Rn.12939
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID s128460

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Vamp2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_012663.2 NP_036795.1 2 151
XM_006246593.2 XP_006246655.1 2 569
GenBank mRNA AJ133104.1 - 2 135
BC074003.1 - 2 151 7097683
M24105.1 - 1 89

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Human: AJ225044.

Gene Ontology Categories:

Function(s) Process(es)

protein complex assembly
vesicle fusion
neurotransmitter secretion
response to glucose
protein transport
synaptic vesicle exocytosis
vesicle-mediated transport
calcium ion regulated exocytosis
regulation of exocytosis
cellular response to insulin stimulus
Golgi to plasma membrane protein transport
eosinophil degranulation
neutrophil degranulation
natural killer cell degranulation
long-term synaptic potentiation
regulation of vesicle-mediated transport
membrane fusion
mucus secretion
positive regulation of intracellular protein transport
regulation of histamine secretion by mast cell
SNARE binding
SNAP receptor activity
protein binding
calmodulin binding
phospholipid binding
protein C-terminus binding
myosin binding
syntaxin-1 binding
syntaxin binding
protein complex binding
identical protein binding
calcium-dependent protein binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-67c6c7f55f-2g6mz:80/100.66.128.142:80.
git-commit: 92b42c745e7842b39e6d37af9c4064bd81a92e4d
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header