This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other PMP22 SIRNA Assays ›
Gene Symbol:
Pmp22
Gene Name:
peripheral myelin protein 22
Gene Aliases:
Gas-3
Chromosome Location:
Chr.10: 49538588 - 49568583 on Build Rnor_6.0
UniGene ID:
Rn.1476
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID s128232

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Pmp22
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_017037.1 NP_058733.1 4 492
XM_006246584.2 XP_006246646.1 4 490
XM_006246585.2 XP_006246647.1 4 499
GenBank mRNA M69139.1 - 4 408
S55427.1 - 4 408
X62431.1 - 4 492

Back To Top

More Information


Panther Classification:

Molecular Function -

cytoskeletal protein

Gene Ontology Categories:

Function(s)

cell cycle arrest
peripheral nervous system development
cell death
negative regulation of cell proliferation
negative regulation of neuron projection development
cell differentiation
bleb assembly
myelin assembly
myelination

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0