This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other PLA2G4A SIRNA Assays ›
Gene Symbol:
Pla2g4a LOC100911675
Gene Name:
phospholipase A2, group IVA (cytosolic, calcium-dependent)
cytosolic phospholipase A2-like
Gene Aliases:
Pla2c, Pla2g4, cPLA2
-
Chromosome Location:
Chr.13: 67062252 - 67206688 on Build Rnor_6.0
Chromosome Location:
Chr.13: 66988937 - 67001440 on Build Rnor_6.0
UniGene ID:
Rn.10162
-
Species:
Rat
Species Specific ID (Flybase ID):
-
-
siRNA ID s128212

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Pla2g4a
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_133551.2 NP_598235.2 17 2228
GenBank mRNA BC070940.1 - 17 2228 7099813
S77829.1 - 17 2214
U38376.1 - 17 2229
Gene Symbol: LOC100911675
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_003751248.3 XP_003751296.1 828

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

ovulation from ovarian follicle
luteolysis
aging
positive regulation of cell proliferation
phospholipid catabolic process
response to heat
response to hormone
response to organic substance
response to lithium ion
response to organonitrogen compound
arachidonic acid metabolic process
positive regulation of bone mineralization
positive regulation of vesicle fusion
positive regulation of prostaglandin biosynthetic process
positive regulation of fever generation
response to lipopolysaccharide
response to vitamin D
regulation of cell proliferation
response to hydrogen peroxide
positive regulation of apoptotic process
surfactant homeostasis
icosanoid biosynthetic process
decidualization
arachidonic acid secretion
positive regulation of inflammatory response
response to glucocorticoid
response to calcium ion
response to methylmercury
cellular response to antibiotic
lysophospholipase activity
phospholipase A2 activity
calcium ion binding
calcium-dependent phospholipid binding
histone acetyltransferase binding
calcium-dependent phospholipase A2 activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-7db4fb6694-9hb2s:80/100.66.128.142:80.
git-commit: 672a5a8c50d6b616e949be631a941c86748ade1b
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header