This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other NPPA SIRNA Assays ›
Gene Symbol:
Nppa
Gene Name:
natriuretic peptide A
Gene Aliases:
ANF, ANP, Pnd, RATANF
Chromosome Location:
Chr.5: 164808407 - 164809716 on Build Rnor_6.0
UniGene ID:
Rn.2004
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID s128120

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Nppa
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_012612.2 NP_036744.1 1, 2 195
GenBank mRNA BC158590.1 - 1, 2 185 9036379
CB724799.1 - 1, 2 322
M27498.1 - 1, 2 183
X00665.1 - 1, 2 187
X01118.1 - 1, 2 176

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_008725, BC089615, AK147180.

Gene Ontology Categories:

Function(s) Process(es)

response to hypoxia
negative regulation of systemic arterial blood pressure
cGMP biosynthetic process
receptor guanylyl cyclase signaling pathway
neuropeptide signaling pathway
heart development
female pregnancy
regulation of blood pressure
positive regulation of heart rate
cardiac muscle hypertrophy in response to stress
negative regulation of cell growth
response to insulin
regulation of body fluid levels
regulation of blood vessel size
multicellular organismal water homeostasis
regulation of atrial cardiac muscle cell membrane repolarization
positive regulation of cardiac muscle contraction
cellular response to mechanical stimulus
regulation of high voltage-gated calcium channel activity
positive regulation of delayed rectifier potassium channel activity
regulation of calcium ion transmembrane transport via high voltage-gated calcium channel
positive regulation of potassium ion export across plasma membrane
negative regulation of collecting lymphatic vessel constriction
receptor binding
hormone activity
neuropeptide hormone activity
peptide hormone receptor binding
neuropeptide receptor binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0