This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other GRIN2D SIRNA Assays ›
Gene Symbol:
Grin2d
Gene Name:
glutamate receptor, ionotropic, N-methyl D-aspartate 2D
Gene Aliases:
GluN2D
Chromosome Location:
Chr.1: 101819026 - 101856881 on Build Rnor_6.0
UniGene ID:
Rn.91209
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID s127817

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Grin2d
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_022797.1 NP_073634.1 10 2897
XM_008759316.1 XP_008757538.1 9 2602
XM_008759317.1 XP_008757539.1 9 2602
GenBank mRNA D13213.1 - 9 2191
L31611.1 - 9 2172
L31612.1 - 10 2897
U08260.1 - 10 2243

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

startle response
cation transport
adult locomotory behavior
ion transmembrane transport
regulation of ion transmembrane transport
ionotropic glutamate receptor signaling pathway
synaptic transmission, glutamatergic
detection of mechanical stimulus involved in sensory perception of pain
regulation of sensory perception of pain
excitatory postsynaptic potential
cation transmembrane transport
ionotropic glutamate receptor activity
NMDA glutamate receptor activity
extracellular-glutamate-gated ion channel activity
cation channel activity
protein binding
glutamate binding
voltage-gated cation channel activity
neurotransmitter binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-56696975f8-skfwn:80/100.66.128.142:80.
git-commit: 795bdb1f794b4b81290d0c522fee4b91276a707f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0