This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other GOT1 SIRNA Assays ›
Gene Symbol:
Got1
Gene Name:
glutamic-oxaloacetic transaminase 1, soluble
Gene Aliases:
Aspat, Gaspat, cAspAT, cCAT
Chromosome Location:
Chr.1: 263246248 - 263269762 on Build Rnor_6.0
UniGene ID:
Rn.5819
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID s127794

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Got1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_012571.2 NP_036703.2 8 1138
GenBank mRNA BC061877.1 - 8 1111 5599273
D00252.1 - 8 1095
FN802619.1 - 2 183
FQ219727.1 - 5 626
J04171.1 - 8 1079

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_010324, AK147934, AK147807, AK146445, J02623, and others.

Gene Ontology Categories:

Function(s) Process(es)

2-oxoglutarate metabolic process
oxaloacetate metabolic process
glycerol biosynthetic process
aspartate metabolic process
aspartate biosynthetic process
aspartate catabolic process
glutamate metabolic process
glutamate catabolic process to aspartate
glutamate catabolic process to 2-oxoglutarate
cellular response to insulin stimulus
dicarboxylic acid metabolic process
response to glucocorticoid
fatty acid homeostasis
L-aspartate:2-oxoglutarate aminotransferase activity
phosphatidylserine decarboxylase activity
pyridoxal phosphate binding
carboxylic acid binding
L-cysteine:2-oxoglutarate aminotransferase activity
L-phenylalanine:2-oxoglutarate aminotransferase activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0