This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other FGG SIRNA Assays ›
Gene Symbol:
Fgg
Gene Name:
fibrinogen gamma chain
Gene Aliases:
-
Chromosome Location:
Chr.2: 181987080 - 181994523 on Build Rnor_6.0
UniGene ID:
Rn.1702
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID s127734

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Fgg
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_012559.2 NP_036691.2 9 1253
XM_006232525.1 XP_006232587.1 9 1407
GenBank mRNA BC078893.1 - 9 1253 7122366
FQ209468.1 - 5 726
FQ210959.1 - 3 511
FQ219408.1 - 9 1278
FQ219511.1 - 9 1275
FQ219541.1 - 9 1274
FQ219604.1 - 9 1277
FQ229132.1 - 9 1280
J00734.1 - 9 1238
J00735.1 - 2 147

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_133862, AK013887, BC019506, BC019828, AK149448, and others.

Gene Ontology Categories:

Function(s) Process(es)

inflammatory response
cell-matrix adhesion
signal transduction
positive regulation of heterotypic cell-cell adhesion
cellular protein complex assembly
positive regulation of vasoconstriction
positive regulation of exocytosis
positive regulation of protein secretion
protein polymerization
response to calcium ion
positive regulation of ERK1 and ERK2 cascade
platelet aggregation
positive regulation of peptide hormone secretion
negative regulation of extrinsic apoptotic signaling pathway via death domain receptors
negative regulation of endothelial cell apoptotic process
molecular_function
receptor binding
structural molecule activity
protein binding, bridging
metal ion binding
cell adhesion molecule binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-7cdd86b58d-kqgnb:80/100.66.128.142:80.
git-commit: 00e1f9c870be1646ae16b59a7cca7819e45b49ab
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0