This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other AR SIRNA Assays ›
Gene Symbol:
Ar
Gene Name:
androgen receptor
Gene Aliases:
Andr, Tfm
Chromosome Location:
Chr.X: 67656253 - 67828998 on Build Rnor_6.0
UniGene ID:
Rn.9813
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID s127466

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Ar
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_012502.1 NP_036634.1 3 2798
GenBank mRNA M20133.1 - 3 2798
M23264.1 - 3 1837

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_013476, X59592, X59411, M37890, AK133647, and others.

Gene Ontology Categories:

Function(s) Process(es)

in utero embryonic development
regulation of systemic arterial blood pressure
epithelial cell morphogenesis
transcription, DNA-templated
regulation of transcription, DNA-templated
regulation of transcription from RNA polymerase II promoter
spermatogenesis
single fertilization
positive regulation of cell proliferation
male gonad development
fertilization
cellular process
regulation of gene expression
male somatic sex determination
androgen receptor signaling pathway
intracellular receptor signaling pathway
response to insulin
positive regulation of intracellular estrogen receptor signaling pathway
Leydig cell differentiation
multicellular organism growth
positive regulation of phosphorylation
steroid hormone mediated signaling pathway
positive regulation of MAPK cascade
positive regulation of insulin-like growth factor receptor signaling pathway
negative regulation of integrin biosynthetic process
positive regulation of integrin biosynthetic process
positive regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
positive regulation of transcription from RNA polymerase III promoter
male sex differentiation
reproductive structure development
regulation of developmental growth
organ formation
male genitalia morphogenesis
negative regulation of epithelial cell proliferation
regulation of catalytic activity
positive regulation of NF-kappaB transcription factor activity
protein oligomerization
activation of prostate induction by androgen receptor signaling pathway
morphogenesis of an epithelial fold
lateral sprouting involved in mammary gland duct morphogenesis
regulation of prostatic bud formation
prostate gland growth
prostate gland epithelium morphogenesis
epithelial cell differentiation involved in prostate gland development
tertiary branching involved in mammary gland duct morphogenesis
mammary gland alveolus development
reproductive system development
cellular response to steroid hormone stimulus
seminiferous tubule development
regulation of establishment of protein localization to plasma membrane
negative regulation of extrinsic apoptotic signaling pathway
RNA polymerase II core promoter proximal region sequence-specific DNA binding
transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
RNA polymerase II transcription factor binding
DNA binding
chromatin binding
transcription factor activity, sequence-specific DNA binding
RNA polymerase II transcription factor activity, ligand-activated sequence-specific DNA binding
androgen receptor activity
receptor binding
androgen binding
protein binding
beta-catenin binding
transcription factor binding
zinc ion binding
enzyme binding
protein domain specific binding
sequence-specific DNA binding
transcription regulatory region DNA binding
androgen receptor binding
POU domain binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0