This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other ALDOC SIRNA Assays ›
Gene Symbol:
Aldoc
Gene Name:
aldolase C, fructose-bisphosphate
Gene Aliases:
ALDCAA, F16dip7, RATALDCAA
Chromosome Location:
Chr.10: 65586504 - 65590093 on Build Rnor_6.0
UniGene ID:
Rn.11211
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID s127445

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Aldoc
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_012497.1 NP_036629.1 2 164
GenBank mRNA BC059154.1 - 2 139
BC099749.1 - 2 150 5599032
FQ213506.1 - 2 167
FQ214420.1 - 2 190
FQ228893.1 - 2 164
X06984.1 - 2 164

Back To Top

More Information


Panther Classification:

Molecular Function -

aldolase

Gene Ontology Categories:

Function(s) Process(es)

response to hypoxia
glycolytic process
apoptotic process
aging
response to organonitrogen compound
response to organic cyclic compound
fructose 1,6-bisphosphate metabolic process
epithelial cell differentiation
organ regeneration
protein homotetramerization
protein heterotetramerization
fructose-bisphosphate aldolase activity
protein binding
cytoskeletal protein binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0